siRNA Details
Gene Symbol: POU5F1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480888.1 | - | 1 | 292 |
Gene Symbol: POU5F1P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480890.1 | - | 1 | 291 | ||
HQ122798.1 | - | 1 | 292 | |||
HQ122800.1 | - | 1 | 292 | |||
HQ122804.1 | - | 1 | 292 | |||
HQ122807.1 | - | 1 | 292 | |||
HQ122811.1 | - | 1 | 292 | |||
HQ122858.1 | - | 1 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
