siRNA Details
Gene Symbol: POU5F1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159542.1 | NP_001153014.1 | 1 | 1262 | ||
GenBank mRNA | AF268615.1 | - | 1 | 1007 | ||
AF268618.1 | - | 1 | 1007 | |||
DQ486513.1 | - | 1 | 1262 | |||
GU480835.1 | - | 1 | 1008 | |||
GU480840.1 | - | 1 | 1008 | |||
GU480841.1 | - | 1 | 1008 | |||
GU480842.1 | - | 1 | 1008 | |||
GU480845.1 | - | 1 | 1008 | |||
GU480849.1 | - | 1 | 1010 | |||
GU480850.1 | - | 1 | 1010 | |||
GU480851.1 | - | 1 | 1009 | |||
GU480864.1 | - | 1 | 1008 | |||
GU480868.1 | - | 1 | 1008 | |||
GU480878.1 | - | 1 | 1008 | |||
GU480880.1 | - | 1 | 1008 | |||
GU480883.1 | - | 1 | 1008 | |||
GU480884.1 | - | 1 | 1007 | |||
GU480885.1 | - | 1 | 1009 | |||
GU480886.1 | - | 1 | 1008 | |||
GU480889.1 | - | 1 | 1008 | |||
GU480896.1 | - | 1 | 1008 | |||
GU480899.1 | - | 1 | 1009 | |||
GU480902.1 | - | 1 | 1008 | |||
GU480905.1 | - | 1 | 1008 | |||
GU480906.1 | - | 1 | 1008 | |||
GU480908.1 | - | 1 | 1008 | |||
GU480910.1 | - | 1 | 1010 | |||
GU480911.1 | - | 1 | 1008 | |||
GU480912.1 | - | 1 | 1008 | |||
GU480913.1 | - | 1 | 1008 | |||
HQ122710.1 | - | 1 | 1008 | |||
HQ122711.1 | - | 1 | 1008 | |||
HQ122712.1 | - | 1 | 1008 | |||
HQ122713.1 | - | 1 | 1008 | |||
HQ122714.1 | - | 1 | 1008 | |||
HQ122715.1 | - | 1 | 1007 | |||
HQ122716.1 | - | 1 | 1008 | |||
HQ122717.1 | - | 1 | 1008 | |||
HQ122718.1 | - | 1 | 1008 | |||
HQ122719.1 | - | 1 | 1008 | |||
HQ122720.1 | - | 1 | 1009 | |||
HQ122721.1 | - | 1 | 1008 | |||
HQ122749.1 | - | 1 | 1008 | |||
HQ122750.1 | - | 1 | 1008 | |||
HQ122751.1 | - | 1 | 1008 | |||
HQ122752.1 | - | 1 | 1008 | |||
HQ122781.1 | - | 1 | 1008 | |||
HQ122782.1 | - | 1 | 1008 | |||
HQ122783.1 | - | 1 | 1008 | |||
HQ122784.1 | - | 1 | 1008 | |||
HQ122785.1 | - | 1 | 1008 | |||
HQ122786.1 | - | 1 | 1008 | |||
HQ122787.1 | - | 1 | 1008 | |||
HQ122788.1 | - | 1 | 1008 | |||
HQ122789.1 | - | 1 | 1008 | |||
HQ122790.1 | - | 1 | 1008 | |||
HQ122816.1 | - | 1 | 1008 | |||
HQ122817.1 | - | 1 | 1008 | |||
HQ122818.1 | - | 1 | 1008 | |||
HQ122819.1 | - | 1 | 1008 | |||
HQ122820.1 | - | 1 | 1008 | |||
HQ122821.1 | - | 1 | 1008 | |||
HQ122822.1 | - | 1 | 1008 | |||
HQ122823.1 | - | 1 | 1008 | |||
HQ122824.1 | - | 1 | 1008 | |||
HQ122825.1 | - | 1 | 1008 | |||
HQ122826.1 | - | 1 | 1008 | |||
HQ122827.1 | - | 1 | 1008 | |||
HQ122841.1 | - | 1 | 1008 | |||
HQ122842.1 | - | 1 | 1008 | |||
HQ122843.1 | - | 1 | 1008 | |||
HQ122844.1 | - | 1 | 1008 | |||
HQ122845.1 | - | 1 | 1008 | |||
HQ122846.1 | - | 1 | 1008 | |||
HQ122847.1 | - | 1 | 1008 | |||
HQ122848.1 | - | 1 | 1008 | |||
HQ122849.1 | - | 1 | 1008 | |||
HQ122850.1 | - | 1 | 1009 | |||
HQ122851.1 | - | 1 | 1008 | |||
HQ122852.1 | - | 1 | 1008 | |||
HQ122853.1 | - | 1 | 1008 | |||
HQ122854.1 | - | 1 | 1008 | |||
HQ122855.1 | - | 1 | 1008 | |||
HQ122856.1 | - | 1 | 1008 | |||
HQ122857.1 | - | 1 | 1008 | |||
HQ122863.1 | - | 1 | 1007 | |||
HQ122864.1 | - | 1 | 1008 | |||
HQ122865.1 | - | 1 | 1008 | |||
HQ122866.1 | - | 1 | 1008 | |||
HQ122867.1 | - | 1 | 1008 | |||
HQ122868.1 | - | 1 | 1008 | |||
HQ122869.1 | - | 1 | 1008 | |||
HQ122870.1 | - | 1 | 1008 | |||
HQ122871.1 | - | 1 | 1008 | |||
HQ122872.1 | - | 1 | 1007 | |||
HQ122873.1 | - | 1 | 1007 | |||
HQ122874.1 | - | 1 | 1008 | |||
HQ122876.1 | - | 1 | 1008 | |||
HQ122877.1 | - | 1 | 1008 | |||
HQ122878.1 | - | 1 | 1008 | |||
HQ122879.1 | - | 1 | 1008 | |||
HQ122880.1 | - | 1 | 1008 | |||
HQ122881.1 | - | 1 | 1008 | |||
HQ122882.1 | - | 1 | 1008 | |||
HQ122883.1 | - | 1 | 1008 | |||
HQ122884.1 | - | 1 | 1008 | |||
HQ122885.1 | - | 1 | 1008 | |||
HQ122886.1 | - | 1 | 1008 | |||
HQ122887.1 | - | 1 | 1008 | |||
HQ122891.1 | - | 1 | 1008 | |||
HQ122892.1 | - | 1 | 1008 | |||
HQ122893.1 | - | 1 | 1008 | |||
HQ122894.1 | - | 1 | 1008 | |||
HQ122895.1 | - | 1 | 1008 | |||
HQ122896.1 | - | 1 | 1008 | |||
HQ122897.1 | - | 1 | 1008 | |||
HQ122898.1 | - | 1 | 1008 | |||
HQ122899.1 | - | 1 | 1008 | |||
HQ122900.1 | - | 1 | 1008 | |||
HQ122901.1 | - | 1 | 1009 | |||
HQ122902.1 | - | 1 | 1008 | |||
HQ122903.1 | - | 1 | 1008 |
Gene Symbol: POU5F1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480897.1 | - | 1 | 1009 |
Gene Symbol: POU5F1P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480856.1 | - | 1 | 1012 | ||
HQ122904.1 | - | 1 | 1012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
