siRNA Details
Gene Symbol: TUBA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207312.2 | NP_997195.1 | 4 | 1027 | ||
GenBank mRNA | AK131043.1 | - | 4 | 1005 | ||
BC057811.1 | - | 4 | 1027 | 5298532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
