siRNA Details
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 989 | |||
NM_001302371.1 | NP_001289300.1 | 989 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 6 | 954 | ||
XM_011509395.2 | XP_011507697.1 | 9 | 1454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
