siRNA Details
Gene Symbol: RRP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015703.4 | NP_056518.2 | 7 | 867 | ||
GenBank mRNA | AK092492.1 | - | 5 | 1393 | ||
AK314650.1 | - | 7 | 833 | |||
AY039240.1 | - | 6 | 739 | |||
BC031838.1 | - | 6 | 782 | |||
BC035992.1 | - | 7 | 846 | |||
BC041639.1 | - | 7 | 825 | |||
BC042335.1 | - | 7 | 828 | |||
BC063537.1 | - | 3 | 363 | |||
BC073834.1 | - | 7 | 840 | 5533064 | ||
BC121118.1 | - | 7 | 850 | 40120513 | ||
BC121119.1 | - | 7 | 850 | 40120514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
