siRNA Details
Gene Symbol: FKBP9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003949.1 | 6 | 903 | |||
NR_027339.1 | 4 | 1007 | ||||
NR_027340.1 | 3 | 636 | ||||
NR_027342.1 | 4 | 917 | ||||
GenBank mRNA | BC011872.2 | - | 4 | 917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
