siRNA Details
Gene Symbol: SLC35G3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152462.2 | NP_689675.1 | 1 | 161 | ||
GenBank mRNA | AK097473.1 | - | 1 | 126 | ||
BC136422.1 | - | 1 | 161 | 9020411 | ||
BC140700.1 | - | 1 | 161 | 9020410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
