siRNA Details
Gene Symbol: POTEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277304.1 | NP_001264233.1 | 11 | 1653 | ||
NR_102391.1 | 9 | 1444 |
Gene Symbol: LOC102723502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724900.3 | XP_006724963.1 | 11 | 1878 |
Gene Symbol: POTEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277303.1 | NP_001264232.1 | 11 | 1653 | ||
NR_102390.1 | 9 | 1444 | ||||
GenBank mRNA | BC101007.1 | - | 9 | 1444 | 40006489 |
Gene Symbol: POTEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207355.3 | NP_997238.2 | 11 | 2102 | ||
GenBank mRNA | AY465170.1 | - | 12 | 1672 | ||
AY465173.1 | - | 11 | 1779 | |||
BC101005.1 | - | 11 | 1653 | 40006483 | ||
BC101006.1 | - | 11 | 1764 | 40006485 | ||
BC127123.1 | - | 11 | 1764 | 40006490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
