siRNA Details
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 1 | 149 | ||
GenBank mRNA | AK291587.1 | - | 1 | 68 | ||
AU134786.2 | - | 1 | 149 | |||
BC002717.2 | - | 1 | 42 | 3631916 | ||
BC005921.1 | - | 1 | 61 | 4043768 | ||
BC020756.1 | - | 1 | 117 | 4769769 | ||
BC057768.1 | - | 1 | 61 | 30337725 | ||
BC062775.1 | - | 1 | 65 | 4604848 | ||
BG436049.1 | - | 1 | 62 | |||
J00118.1 | - | 1 | 28 | |||
M35419.1 | - | 1 | 162 |
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK300190.1 | - | 1 | 61 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
