siRNA Details
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 13 | 1546 | ||
NM_173614.2 | NP_775885.1 | 13 | 1546 | |||
XM_017023156.1 | XP_016878645.1 | 13 | 1593 | |||
GenBank mRNA | AK225803.1 | - | 13 | 1516 | ||
AL832855.1 | - | 13 | 1540 | |||
BC028389.1 | - | 12 | 1406 | 4824724 | ||
BC041131.1 | - | 13 | 1520 | 5757234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
