siRNA Details
Gene Symbol: DGCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005675.4 | NP_005666.2 | 2, 3 | 416 | ||
XM_011530394.1 | XP_011528696.1 | 2, 3 | 284 | |||
GenBank mRNA | AF228707.1 | - | 2, 3 | 360 | ||
AK315115.1 | - | 2, 3 | 321 | |||
BC047039.1 | - | 2, 3 | 309 | 5229172 | ||
BQ639188.1 | - | 2, 3 | 416 | |||
CR456434.1 | - | 2, 3 | 286 | |||
KJ534820.1 | - | 2, 3 | 264 | |||
KJ534971.1 | - | 2, 3 | 264 | |||
KJ535003.1 | - | 2, 3 | 264 | |||
KJ535027.1 | - | 2, 3 | 250 | |||
X96484.1 | - | 2, 3 | 278 |
Gene Symbol: LOC102724770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724934.1 | XP_006724997.1 | 2, 3 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
