siRNA Details
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000319.1 | - | 2 | 608 | ||
AK024774.1 | - | 1 | 307 | |||
AK300522.1 | - | 3 | 628 | |||
AK311305.1 | - | 4 | 672 | |||
BC085018.1 | - | 3 | 857 | 6650445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
