siRNA Details
Gene Symbol: GPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321525.1 | NP_001308454.1 | 21 | 2651 | ||
NM_001321526.1 | NP_001308455.1 | 21 | 2741 | |||
NM_001321527.1 | NP_001308456.1 | 21 | 2705 | |||
NM_001321528.1 | NP_001308457.1 | 21 | 2695 | |||
NM_001321529.1 | NP_001308458.1 | 20 | 2528 | |||
NM_001321530.1 | NP_001308459.1 | 20 | 2420 | |||
NM_001321531.1 | NP_001308460.1 | 20 | 2441 | |||
NM_207328.3 | NP_997211.2 | 21 | 2723 | |||
XM_005263888.3 | XP_005263945.1 | 19 | 2145 | |||
XM_006712311.3 | XP_006712374.1 | 20 | 2295 | |||
XM_006712313.3 | XP_006712376.1 | 19 | 2163 | |||
XM_017003424.1 | XP_016858913.1 | 21 | 2355 | |||
XM_017003426.1 | XP_016858915.1 | 18 | 2085 | |||
GenBank mRNA | AK122580.1 | - | 11 | 4147 | ||
AK122734.1 | - | 13 | 1463 | |||
AK131194.1 | - | 20 | 2441 | |||
AK131197.1 | - | 15 | 1760 | |||
AK298131.1 | - | 20 | 2100 | |||
AK302887.1 | - | 19 | 3455 | |||
BC068596.1 | - | 21 | 2394 | 4837765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
