siRNA Details
Gene Symbol: AQP12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198998.2 | NP_945349.1 | 2 | 349 | ||
GenBank mRNA | AB040748.1 | - | 2 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
