siRNA Details
Gene Symbol: RGPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144013.1 | NP_001137485.1 | 20 | 3493 | ||
XM_017004738.1 | XP_016860227.1 | 21 | 3542 | |||
XM_017004739.1 | XP_016860228.1 | 21 | 3542 | |||
GenBank mRNA | AK310529.1 | - | 6 | 1272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
