siRNA Details
Gene Symbol: DPPA3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY230136.1 | - | 1 | 245 | ||
BC062480.1 | - | 1 | 438 | 30398001 | ||
BC125145.1 | - | 1 | 257 | 40120129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
