siRNA Details
Gene Symbol: KRT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000224.2 | NP_000215.1 | 7 | 1409 | ||
NM_199187.1 | NP_954657.1 | 8 | 1363 | |||
GenBank mRNA | AK129587.1 | - | 8 | 1285 | ||
AK223093.1 | - | 7 | 1351 | |||
AY762101.1 | - | 7 | 1298 | |||
BC000180.2 | - | 7 | 1335 | 2984862 | ||
BC000698.2 | - | 7 | 1336 | 3349315 | ||
BC004253.1 | - | 7 | 1320 | 3631540 | ||
BC008636.1 | - | 7 | 1333 | 3463780 | ||
BC020982.1 | - | 7 | 1343 | 3850724 | ||
BC072017.1 | - | 7 | 1339 | 6251574 | ||
X12876.1 | - | 5 | 700 | |||
X12881.1 | - | 7 | 1340 | |||
X12883.1 | - | 7 | 1276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
