siRNA Details
Gene Symbol: VCX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF167079.1 | - | 2 | 273 | ||
BC104167.1 | - | 1 | 82 | 40030354 | ||
BC104168.1 | - | 1 | 82 | 40030355 |
Gene Symbol: VCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013452.2 | NP_038480.2 | 2 | 185 | ||
XM_011545490.2 | XP_011543792.1 | 1 | 77 | |||
GenBank mRNA | AF167081.2 | - | 2 | 185 | ||
BC126903.1 | - | 1 | 82 | 40030361 |
Gene Symbol: VCX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC126902.1 | - | 1 | 82 | 40030357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
