siRNA Details
Gene Symbol: VCX3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001888.3 | NP_001001888.3 | 3 | 936 | ||
GenBank mRNA | AF167080.2 | - | 4 | 846 | ||
BC098143.1 | - | 3 | 378 | 40014420 |
Gene Symbol: VCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013452.2 | NP_038480.2 | 3 | 758 | ||
GenBank mRNA | AF159129.1 | - | 2 | 565 | ||
AF167081.2 | - | 3 | 758 | |||
BC098123.1 | - | 3 | 564 | 40002642 | ||
BC126903.1 | - | 2 | 655 | 40030361 |
Gene Symbol: VCX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC126902.1 | - | 3 | 595 | 40030357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
