siRNA Details
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122842.1 | - | 6 | 3877 | ||
BC041705.1 | - | 2 | 1094 | |||
BC053641.1 | - | 2 | 904 | |||
BC063619.1 | - | 1 | 1242 | |||
BC083493.1 | - | 1 | 1184 |
Gene Symbol: BCRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037566.1 | 6 | 2018 |
Gene Symbol: BCRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024494.1 | 7 | 1110 |
Gene Symbol: LOC102724751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030165.1 | XP_016885654.1 | 3 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
