siRNA Details
Gene Symbol: RFPL4AL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277397.1 | NP_001264326.1 | 2 | 299 | ||
XM_011527259.1 | XP_011525561.1 | 3 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
