siRNA Details
Gene Symbol: RGPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182588.2 | NP_872394.2 | 20 | 3840 | ||
XM_011511017.2 | XP_011509319.1 | 20 | 3787 | |||
XM_011511018.2 | XP_011509320.1 | 20 | 3790 | |||
XM_011511019.2 | XP_011509321.1 | 20 | 3786 | |||
XM_011511020.2 | XP_011509322.1 | 20 | 3786 | |||
GenBank mRNA | BX537861.1 | - | 15 | 4739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
