siRNA Details
Gene Symbol: RGPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182588.2 | NP_872394.2 | 12, 13 | 1845 | ||
XM_011511017.2 | XP_011509319.1 | 12, 13 | 1792 | |||
XM_011511018.2 | XP_011509320.1 | 12, 13 | 1795 | |||
XM_011511019.2 | XP_011509321.1 | 12, 13 | 1791 | |||
XM_011511020.2 | XP_011509322.1 | 12, 13 | 1791 | |||
XM_017003898.1 | XP_016859387.1 | 12, 13 | 1856 | |||
GenBank mRNA | AK302921.1 | - | 8, 9 | 1153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
