siRNA Details
Gene Symbol: C2orf27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013310.3 | NP_037442.3 | 1 | 62 | ||
GenBank mRNA | AF038169.1 | - | 1 | 62 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
