siRNA Details
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 4 | 2140 | ||
NM_001320149.1 | NP_001307078.1 | 4 | 1613 | |||
XM_011542018.1 | XP_011540320.1 | 5 | 2548 | |||
GenBank mRNA | BC046199.1 | - | 4 | 1613 | 6064384 |
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123168.2 | NP_001116640.1 | 4 | 2163 | ||
NM_001317901.1 | NP_001304830.1 | 4 | 1383 | |||
NR_134239.1 | 5 | 1443 | ||||
XM_006711526.3 | XP_006711589.1 | 5 | 2137 | |||
XM_011509965.2 | XP_011508267.1 | 5 | 2046 | |||
XM_011509967.1 | XP_011508269.1 | 4 | 2058 | |||
XM_017002233.1 | XP_016857722.1 | 5 | 2043 | |||
GenBank mRNA | BC035696.1 | - | 4 | 1730 | 5541342 | |
DQ786198.1 | - | 1 | 477 |
Gene Symbol: FAM72C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287385.1 | NP_001274314.1 | 4 | 2178 | ||
XM_011509723.2 | XP_011508025.1 | 5 | 2231 | |||
XM_017001682.1 | XP_016857171.1 | 5 | 2428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
