siRNA Details
Gene Symbol: SLC35E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199787.1 | NP_001186716.1 | 1 | 188 | ||
NM_182838.2 | NP_878258.1 | 1 | 188 | |||
GenBank mRNA | AL049665.1 | - | 1 | 181 | ||
DB461679.1 | - | 1 | 189 |
Gene Symbol: SLC35E2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC110653.1 | - | 1 | 244 | 4392812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
