siRNA Details
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005362.3 | NP_005353.1 | 3 | 1132 | ||
XM_005274676.3 | XP_005274733.1 | 3 | 1206 | |||
XM_006724818.2 | XP_006724881.1 | 3 | 1288 | |||
XM_011531160.2 | XP_011529462.1 | 3 | 1112 | |||
XM_011531161.2 | XP_011529463.1 | 3 | 1537 | |||
GenBank mRNA | AK223519.1 | - | 1 | 959 | ||
AK292384.1 | - | 3 | 1132 | |||
BC000340.2 | - | 3 | 1097 | 2822978 | ||
BC005963.1 | - | 3 | 1132 | 4064879 | ||
BC011744.2 | - | 2 | 1058 | 3345801 | ||
BC016803.1 | - | 3 | 1094 | 4073332 | ||
BC017389.1 | - | 3 | 1089 | 4047542 | ||
CR541767.1 | - | 1 | 923 | |||
CR541774.1 | - | 1 | 923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
