siRNA Details
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 7000 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 6066 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 6867 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 6554 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 7129 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 7393 | |||
NM_003879.5 | NP_003870.4 | 10 | 6972 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 7867 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 6836 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 6915 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 7802 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 9819 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 7675 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 7013 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 6638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
