siRNA Details
Gene Symbol: BMS1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027293.2 | 6 | 628 | |||
GenBank mRNA | AK090629.1 | - | 6 | 568 | ||
BX641141.1 | - | 5 | 2124 |
Gene Symbol: BMS1P15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC107108.2 | - | 3 | 366 |
Gene Symbol: LOC101929959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015376.1 | XP_016870865.1 | 7 | 933 | ||
XM_017015377.1 | XP_016870866.1 | 7 | 933 | |||
XM_017015379.1 | XP_016870868.1 | 7 | 892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
