siRNA Details
Gene Symbol: TCAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130025.1 | NP_001123497.1 | 4, 5 | 1803 | ||
NM_001130026.2 | NP_001123498.2 | 4, 5 | 1432 | |||
NM_173678.2 | NP_775949.2 | 4, 5 | 1789 | |||
XM_006715927.3 | XP_006715990.1 | 3, 4 | 2664 | |||
XM_006715928.3 | XP_006715991.1 | 3, 4 | 2664 | |||
XM_011516059.2 | XP_011514361.1 | 3, 4 | 1252 | |||
GenBank mRNA | AK098041.1 | - | 3, 4 | 1163 | ||
AK296318.1 | - | 4, 5 | 1408 | |||
AY167570.1 | - | 4, 5 | 1786 | |||
BC113530.1 | - | 4, 5 | 1789 | 8322582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
