siRNA Details
Gene Symbol: NUDT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018159.3 | NP_060629.2 | 1 | 364 | ||
GenBank mRNA | AK001490.1 | - | 1 | 251 | ||
BC009942.2 | - | 1 | 318 | 4303422 | ||
BX419815.2 | - | 1 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
