siRNA Details
Gene Symbol: NOTCH2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203458.4 | NP_982283.2 | 1 | 99 | ||
GenBank mRNA | BC010154.2 | - | 1 | 93 | 3623163 | |
BC019835.1 | - | 1 | 70 | |||
BC065219.1 | - | 1 | 57 | 5724338 | ||
BC084572.1 | - | 1 | 53 | |||
BM792637.1 | - | 1 | 99 |
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG718124.1 | - | 1 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
