siRNA Details
Gene Symbol: LOC100996717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003047.1 | XP_016858536.1 | 3 | 272 | ||
XM_017003048.1 | XP_016858537.1 | 3 | 6967 | |||
XM_017003049.1 | XP_016858538.1 | 3 | 284 | |||
XM_017003050.1 | XP_016858539.1 | 3 | 518 | |||
XM_017003051.1 | XP_016858540.1 | 3 | 517 | |||
XM_017003052.1 | XP_016858541.1 | 4 | 418 |
Gene Symbol: LOC101929796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542531.2 | XP_011540833.1 | 3 | 547 | ||
XM_017003012.1 | XP_016858501.1 | 3 | 547 | |||
XM_017003013.1 | XP_016858502.1 | 3 | 547 | |||
XM_017003014.1 | XP_016858503.1 | 3 | 547 |
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF315356.1 | - | 3 | 317 | ||
BG718124.1 | - | 3 | 560 |
Gene Symbol: NOTCH2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203458.4 | NP_982283.2 | 3 | 556 | ||
GenBank mRNA | AK075065.1 | - | 3 | 534 | ||
BC010154.2 | - | 3 | 550 | 3623163 | ||
BC019835.1 | - | 3 | 527 | |||
BC065219.1 | - | 3 | 508 | 5724338 | ||
BC084572.1 | - | 3 | 510 | |||
BC090943.1 | - | 3 | 526 | 5404327 | ||
BM792637.1 | - | 3 | 548 | |||
BX537434.1 | - | 4 | 635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
