siRNA Details
Gene Symbol: NOTCH2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203458.4 | NP_982283.2 | 4 | 688 | ||
GenBank mRNA | AK075065.1 | - | 4 | 666 | ||
BC010154.2 | - | 4 | 682 | 3623163 | ||
BC019835.1 | - | 4 | 659 | |||
BC065219.1 | - | 4 | 640 | 5724338 | ||
BC084572.1 | - | 4 | 642 | |||
BC090943.1 | - | 4 | 658 | 5404327 | ||
BX537434.1 | - | 5 | 767 |
Gene Symbol: LOC100996717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003047.1 | XP_016858536.1 | 4 | 404 | ||
XM_017003048.1 | XP_016858537.1 | 4 | 7099 | |||
XM_017003049.1 | XP_016858538.1 | 4 | 416 | |||
XM_017003050.1 | XP_016858539.1 | 4 | 650 | |||
XM_017003051.1 | XP_016858540.1 | 4 | 649 | |||
XM_017003052.1 | XP_016858541.1 | 5 | 550 |
Gene Symbol: LOC100996763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711700.3 | XP_006711763.1 | 4 | 597 | ||
XM_006711703.3 | XP_006711766.1 | 4 | 659 | |||
XM_011510256.2 | XP_011508558.1 | 4 | 1384 |
Gene Symbol: LOC101929796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542531.2 | XP_011540833.1 | 4 | 679 | ||
XM_017003012.1 | XP_016858501.1 | 4 | 679 | |||
XM_017003013.1 | XP_016858502.1 | 4 | 679 | |||
XM_017003014.1 | XP_016858503.1 | 4 | 679 |
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF315356.1 | - | 4 | 449 | ||
BG718124.1 | - | 4 | 694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
