siRNA Details
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 7 | 1152 | ||
NM_001322855.1 | NP_001309784.1 | 8 | 1294 | |||
NM_001322856.1 | NP_001309785.1 | 8 | 1251 | |||
NM_001322857.1 | NP_001309786.1 | 7 | 1072 | |||
NM_001322858.1 | NP_001309787.1 | 8 | 1169 | |||
NM_001322859.1 | NP_001309788.1 | 5 | 778 | |||
NM_001322860.1 | NP_001309789.1 | 4 | 718 | |||
NM_001322861.1 | NP_001309790.1 | 4 | 428 | |||
NM_001322862.1 | NP_001309791.1 | 8 | 1129 | |||
NM_001322863.1 | NP_001309792.1 | 7 | 1101 | |||
NM_001322864.1 | NP_001309793.1 | 6 | 669 | |||
NM_001322865.1 | NP_001309794.1 | 7 | 1019 | |||
NM_001322866.1 | NP_001309795.1 | 7 | 992 | |||
NM_001322868.1 | NP_001309797.1 | 7 | 1019 | |||
NM_020448.4 | NP_065181.1 | 7 | 1014 | |||
NR_136507.1 | 4 | 718 | ||||
NR_136508.1 | 7 | 1019 | ||||
XM_011541808.2 | XP_011540110.1 | 6 | 929 | |||
XM_017001868.1 | XP_016857357.1 | 6 | 664 | |||
XM_017001869.1 | XP_016857358.1 | 7 | 999 | |||
XM_017001870.1 | XP_016857359.1 | 8 | 1264 | |||
GenBank mRNA | AK312433.1 | - | 7 | 968 | ||
BC001265.1 | - | 7 | 956 | 3452234 | ||
BC063583.1 | - | 7 | 944 | 4386115 | ||
BX640883.1 | - | 8 | 1076 | |||
U90918.1 | - | 7 | 905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
