siRNA Details
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 6 | 955 | ||
NM_001322855.1 | NP_001309784.1 | 7 | 1097 | |||
NM_001322856.1 | NP_001309785.1 | 7 | 1054 | |||
NM_001322857.1 | NP_001309786.1 | 6 | 875 | |||
NM_001322858.1 | NP_001309787.1 | 7 | 972 | |||
NM_001322859.1 | NP_001309788.1 | 4 | 581 | |||
NM_001322860.1 | NP_001309789.1 | 3 | 521 | |||
NM_001322861.1 | NP_001309790.1 | 3 | 231 | |||
NM_001322862.1 | NP_001309791.1 | 7 | 932 | |||
NM_001322863.1 | NP_001309792.1 | 6 | 904 | |||
NM_001322864.1 | NP_001309793.1 | 5 | 472 | |||
NM_001322865.1 | NP_001309794.1 | 6 | 822 | |||
NM_001322866.1 | NP_001309795.1 | 6 | 795 | |||
NM_001322868.1 | NP_001309797.1 | 6 | 822 | |||
NM_020448.4 | NP_065181.1 | 6 | 817 | |||
NR_136506.1 | 6 | 822 | ||||
NR_136507.1 | 3 | 521 | ||||
NR_136508.1 | 6 | 822 | ||||
XM_011541808.2 | XP_011540110.1 | 5 | 732 | |||
XM_017001868.1 | XP_016857357.1 | 5 | 467 | |||
XM_017001869.1 | XP_016857358.1 | 6 | 802 | |||
XM_017001870.1 | XP_016857359.1 | 7 | 1067 | |||
GenBank mRNA | AK312433.1 | - | 6 | 771 | ||
BC001265.1 | - | 6 | 759 | 3452234 | ||
BC063583.1 | - | 6 | 747 | 4386115 | ||
BX640883.1 | - | 7 | 879 | |||
U90918.1 | - | 6 | 708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
