siRNA Details
Gene Symbol: CRAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020825.3 | NP_065876.3 | 11 | 2493 | ||
GenBank mRNA | AB037847.2 | - | 9 | 1886 | ||
BC137172.1 | - | 10 | 2043 | 9021169 | ||
BC137173.1 | - | 10 | 2043 | 9021170 | ||
BC152477.1 | - | 9 | 1886 | 8862615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
