siRNA Details
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000410.3 | NP_000401.1 | 2 | 312 | ||
NM_001300749.1 | NP_001287678.1 | 2 | 312 | |||
NM_139003.2 | NP_620572.1 | 2 | 312 | |||
NM_139004.2 | NP_620573.1 | 2 | 312 | |||
NM_139006.2 | NP_620575.1 | 2 | 312 | |||
XM_011514543.2 | XP_011512845.1 | 2 | 274 | |||
GenBank mRNA | AF079407.1 | - | 2 | 188 | ||
AF115264.1 | - | 2 | 158 | |||
AF115265.1 | - | 2 | 163 | |||
AF144242.1 | - | 2 | 275 | |||
AF149804.1 | - | 2 | 196 | |||
AJ249337.1 | - | 2 | 312 | |||
AK293393.1 | - | 2 | 164 | |||
AK300933.1 | - | 2 | 156 | |||
AK316502.1 | - | 2 | 164 | |||
BC074721.2 | - | 2 | 201 | 30915171 | ||
BC117201.1 | - | 2 | 187 | 40125752 | ||
BC117203.1 | - | 2 | 187 | 40125754 | ||
JN375754.1 | - | 2 | 113 | |||
U60319.1 | - | 2 | 373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
