siRNA Details
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000410.3 | NP_000401.1 | 2 | 336 | ||
NM_001300749.1 | NP_001287678.1 | 2 | 336 | |||
NM_139003.2 | NP_620572.1 | 2 | 336 | |||
NM_139004.2 | NP_620573.1 | 2 | 336 | |||
NM_139006.2 | NP_620575.1 | 2 | 336 | |||
NM_139009.2 | NP_620578.1 | 2 | 267 | |||
XM_011514543.2 | XP_011512845.1 | 2 | 298 | |||
GenBank mRNA | AF079407.1 | - | 2 | 212 | ||
AF115264.1 | - | 2 | 182 | |||
AF115265.1 | - | 2 | 187 | |||
AF144242.1 | - | 2 | 299 | |||
AF149804.1 | - | 2 | 220 | |||
AJ249335.1 | - | 2 | 267 | |||
AJ249337.1 | - | 2 | 336 | |||
AK293393.1 | - | 2 | 188 | |||
AK300933.1 | - | 2 | 180 | |||
AK316502.1 | - | 2 | 188 | |||
BC074721.2 | - | 2 | 225 | 30915171 | ||
BC117201.1 | - | 2 | 211 | 40125752 | ||
BC117203.1 | - | 2 | 211 | 40125754 | ||
JN375754.1 | - | 2 | 137 | |||
U60319.1 | - | 2 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
