siRNA Details
Gene Symbol: BTBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002860.3 | NP_001002860.2 | 5 | 1806 | ||
NM_001289133.1 | NP_001276062.1 | 3 | 538 | |||
XM_011536939.2 | XP_011535241.1 | 5 | 1571 | |||
XM_017021438.1 | XP_016876927.1 | 2 | 362 | |||
GenBank mRNA | AB040958.2 | - | 5 | 1735 | ||
AK021953.1 | - | 3 | 1025 | |||
AK291422.1 | - | 3 | 479 | |||
AL049394.1 | - | 1 | 60 | |||
BX248762.1 | - | 3 | 485 | |||
BX538231.1 | - | 3 | 127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
