siRNA Details
Gene Symbol: BTBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002860.3 | NP_001002860.2 | 6 | 1939 | ||
NM_001289133.1 | NP_001276062.1 | 4 | 671 | |||
XM_011536939.2 | XP_011535241.1 | 6 | 1704 | |||
XM_017021438.1 | XP_016876927.1 | 3 | 495 | |||
GenBank mRNA | AB040958.2 | - | 6 | 1868 | ||
AK021953.1 | - | 4 | 1158 | |||
AK291422.1 | - | 4 | 612 | |||
AK303355.1 | - | 2 | 405 | |||
AL049394.1 | - | 2 | 193 | |||
BX248762.1 | - | 4 | 618 | |||
BX538231.1 | - | 4 | 260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
