siRNA Details
Gene Symbol: RTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283035.1 | NP_001269964.1 | 10 | 1671 | ||
NM_001283036.1 | NP_001269965.1 | 9 | 1578 | |||
NM_001283037.1 | NP_001269966.1 | 8 | 1526 | |||
NM_016407.4 | NP_057491.2 | 9 | 1581 | |||
XM_017027872.1 | XP_016883361.1 | 12 | 1774 | |||
GenBank mRNA | AF117231.1 | - | 9 | 1531 | ||
AF161513.1 | - | 9 | 1541 | |||
AF161518.1 | - | 9 | 1507 | |||
AF212244.1 | - | 9 | 1519 | |||
AK000481.1 | - | 9 | 1511 | |||
AK000586.1 | - | 8 | 1474 | |||
AK026118.1 | - | 10 | 1431 | |||
BC003359.1 | - | 9 | 1508 | 2900332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
