siRNA Details
Gene Symbol: RTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283035.1 | NP_001269964.1 | 9, 10 | 945 | ||
NM_001283036.1 | NP_001269965.1 | 8, 9 | 852 | |||
NM_001283037.1 | NP_001269966.1 | 7, 8 | 800 | |||
NM_016407.4 | NP_057491.2 | 8, 9 | 855 | |||
XM_017027872.1 | XP_016883361.1 | 11, 12 | 1048 | |||
GenBank mRNA | AF117231.1 | - | 8, 9 | 803 | ||
AF161513.1 | - | 8, 9 | 812 | |||
AF161518.1 | - | 8, 9 | 781 | |||
AF212244.1 | - | 8, 9 | 792 | |||
AK000481.1 | - | 8, 9 | 785 | |||
AK000586.1 | - | 7, 8 | 748 | |||
AK026118.1 | - | 8, 9 | 783 | |||
AK225135.1 | - | 8, 9 | 770 | |||
AK293662.1 | - | 8, 9 | 704 | |||
AK302029.1 | - | 9, 10 | 888 | |||
AK314563.1 | - | 8, 9 | 810 | |||
AY320413.1 | - | 2, 3 | 143 | |||
BC003359.1 | - | 8, 9 | 782 | 2900332 | ||
CR983692.1 | - | 7, 8 | 711 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
