siRNA Details
Gene Symbol: BRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009877.2 | NP_001009877.2 | 14 | 1466 | ||
NM_001317951.1 | NP_001304880.1 | 14 | 1818 | |||
NM_023924.4 | NP_076413.3 | 14 | 1639 | |||
NR_134296.1 | 14 | 1975 | ||||
GenBank mRNA | AK023503.1 | - | 14 | 1796 | ||
AK024392.1 | - | 11 | 3819 | |||
AK026830.1 | - | 9 | 770 | |||
AK297573.1 | - | 14 | 1453 | |||
AK299157.1 | - | 14 | 1639 | |||
AL833925.1 | - | 6 | 506 | |||
AY358630.1 | - | 4 | 1162 | |||
BC007217.1 | - | 15 | 1755 | |||
BC041590.1 | - | 12 | 1325 | 5428011 | ||
BX648703.1 | - | 14 | 1977 | |||
DQ248311.1 | - | 10 | 964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
