siRNA Details
Gene Symbol: BRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009877.2 | NP_001009877.2 | 16 | 1973 | ||
NM_001317951.1 | NP_001304880.1 | 16 | 2325 | |||
NM_023924.4 | NP_076413.3 | 16 | 2146 | |||
NR_134296.1 | 17 | 2632 | ||||
GenBank mRNA | AK023503.1 | - | 16 | 2303 | ||
AK024392.1 | - | 13 | 4326 | |||
AK026830.1 | - | 11 | 1357 | |||
AK125518.1 | - | 2 | 1374 | |||
AK297573.1 | - | 16 | 1960 | |||
AK299157.1 | - | 16 | 2146 | |||
AY358630.1 | - | 6 | 1669 | |||
BC007217.1 | - | 19 | 2935 | |||
BC041590.1 | - | 14 | 1832 | 5428011 | ||
BX648703.1 | - | 17 | 2634 | |||
DQ248311.1 | - | 12 | 1471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
