siRNA Details
Gene Symbol: ERCC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000124.3 | NP_000115.1 | 3, 4 | 730 | ||
NM_001277058.1 | NP_001263987.1 | 3, 4 | 730 | |||
NM_001277059.1 | NP_001263988.1 | 3, 4 | 675 | |||
GenBank mRNA | AK291018.1 | - | 3, 4 | 647 | ||
BC034479.1 | - | 3, 4 | 680 | 4822982 | ||
BC127104.1 | - | 3, 4 | 614 | 40124639 | ||
L04791.1 | - | 3, 4 | 633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
