siRNA Details
Gene Symbol: LRRC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020794.2 | NP_065845.1 | 1 | 177 | ||
XM_017001885.1 | XP_016857374.1 | 3 | 524 | |||
XM_017001886.1 | XP_016857375.1 | 3 | 524 | |||
XM_017001887.1 | XP_016857376.1 | 3 | 524 | |||
XM_017001888.1 | XP_016857377.1 | 3 | 524 | |||
XM_017001889.1 | XP_016857378.1 | 3 | 524 | |||
XM_017001890.1 | XP_016857379.1 | 3 | 1771 | |||
XM_017001891.1 | XP_016857380.1 | 4 | 1848 | |||
XM_017001892.1 | XP_016857381.1 | 3 | 524 | |||
XM_017001893.1 | XP_016857382.1 | 3 | 320 | |||
XM_017001894.1 | XP_016857383.1 | 4 | 366 | |||
XM_017001895.1 | XP_016857384.1 | 3 | 525 | |||
GenBank mRNA | AF434715.1 | - | 1 | 177 | ||
AF498274.1 | - | 3 | 451 | |||
AK294206.1 | - | 3 | 628 | |||
BC128989.1 | - | 1 | 167 | 8381584 | ||
CR749629.1 | - | 1 | 127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
