siRNA Details
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159597.2 | NP_001153069.1 | 3 | 326 | ||
NM_001282682.1 | NP_001269611.1 | 3 | 297 | |||
NM_001282687.1 | NP_001269616.1 | 3 | 346 | |||
NM_015677.3 | NP_056492.2 | 3 | 326 | |||
NR_104223.1 | 5 | 1885 | ||||
NR_104224.1 | 4 | 453 | ||||
NR_104225.1 | 3 | 521 | ||||
NR_104226.1 | 4 | 986 | ||||
NR_104227.1 | 3 | 1258 | ||||
GenBank mRNA | AK026507.1 | - | 3 | 261 | ||
AK026586.1 | - | 3 | 521 | |||
AK055801.1 | - | 3 | 985 | |||
AK096440.1 | - | 3 | 836 | |||
AK123829.1 | - | 4 | 1938 | |||
AK292312.1 | - | 3 | 241 | |||
AL050373.1 | - | 3 | 359 | |||
BC008374.1 | - | 3 | 235 | 3996066 | ||
BC008375.1 | - | 3 | 253 | 3996120 | ||
BC030778.1 | - | 4 | 402 | |||
BC034974.1 | - | 4 | 402 | |||
BC043234.1 | - | 5 | 1890 | |||
CR457361.1 | - | 2 | 213 | |||
CX758922.1 | - | 3 | 531 | |||
DA271598.1 | - | 3 | 294 | |||
DB244375.1 | - | 3 | 297 | |||
HY040340.1 | - | 3 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
