siRNA Details
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159597.2 | NP_001153069.1 | 6, 7 | 653 | ||
NM_001282682.1 | NP_001269611.1 | 8, 9 | 758 | |||
NM_001282687.1 | NP_001269616.1 | 8, 9 | 807 | |||
NM_015677.3 | NP_056492.2 | 6, 7 | 653 | |||
NR_104223.1 | 8, 9 | 2212 | ||||
NR_104225.1 | 6, 7 | 848 | ||||
NR_104226.1 | 8, 9 | 1373 | ||||
NR_104227.1 | 8, 9 | 1719 | ||||
GenBank mRNA | AK026507.1 | - | 6, 7 | 588 | ||
AK026586.1 | - | 6, 7 | 848 | |||
AK055801.1 | - | 6, 7 | 1312 | |||
AK096440.1 | - | 8, 9 | 1297 | |||
AK123829.1 | - | 9, 10 | 2451 | |||
AK292312.1 | - | 8, 9 | 702 | |||
AL050373.1 | - | 8, 9 | 820 | |||
BC008374.1 | - | 6, 7 | 562 | 3996066 | ||
BC008375.1 | - | 6, 7 | 580 | 3996120 | ||
BC043234.1 | - | 8, 9 | 2217 | |||
CR457361.1 | - | 5, 6 | 540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
