siRNA Details
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159597.2 | NP_001153069.1 | 9 | 1093 | ||
NM_001282682.1 | NP_001269611.1 | 11 | 1198 | |||
NM_001282687.1 | NP_001269616.1 | 12 | 1304 | |||
NM_015677.3 | NP_056492.2 | 10 | 1150 | |||
NR_104223.1 | 12 | 2709 | ||||
NR_104225.1 | 10 | 1341 | ||||
NR_104226.1 | 11 | 1813 | ||||
NR_104227.1 | 11 | 2635 | ||||
GenBank mRNA | AK026507.1 | - | 9 | 1028 | ||
AK026586.1 | - | 10 | 1341 | |||
AK096440.1 | - | 11 | 2213 | |||
AK123829.1 | - | 12 | 2891 | |||
AK292312.1 | - | 11 | 1142 | |||
AL050373.1 | - | 12 | 1317 | |||
BC008374.1 | - | 10 | 1059 | 3996066 | ||
BC008375.1 | - | 9 | 1020 | 3996120 | ||
BC043234.1 | - | 12 | 2714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
